Learn More
Invitrogen™ C4orf36 Monoclonal Antibody (OTI8B7)

Description
C4orf36 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | C4orf36 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI8B7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | C4orf36 |
| Gene Accession No. | Q96KX1 |
| Gene Alias | C4orf36; chromosome 4 open reading frame 36; uncharacterized protein C4orf36 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.