Learn More
C4orf36 Mouse anti-Human, Clone: OTI2H5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF808367

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
C4orf36 (Chromosome 4 Open Reading Frame 36) is a Protein Coding gene. An important paralog of this gene is ENSG00000285458.Specifications
C4orf36 | |
Monoclonal | |
Unconjugated | |
C4orf36 | |
chromosome 4 open reading frame 36 | |
Mouse | |
Affinity Chromatography | |
RUO | |
132989 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2H5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q96KX1 | |
C4orf36 | |
Full length human recombit protein of human C4orf36 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.