Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C4orf36 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563441
Description
Immunogen sequence: QLTKCTTIKD GLLPSAESIK LEREYEVKRL CKLKCQENTS KEIQLLLRER PAGLRRP Highest antigen sequence identity to the following orthologs: Mouse - 53%, Rat - 51%.
C4orf36 (Chromosome 4 Open Reading Frame 36) is a Protein Coding gene. An important paralog of this gene is ENSG00000285458.
Specifications
C4orf36 | |
Polyclonal | |
Unconjugated | |
C4orf36 | |
C4orf36; chromosome 4 open reading frame 36; uncharacterized protein C4orf36 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
132989 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96KX1 | |
C4orf36 | |
Recombinant protein corresponding to Human C4orf36. Recombinant protein control fragment (Product #RP-100300). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction