Learn More
C4orf42 Mouse anti-Human, Clone: OTI2C6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF503479

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
C4orf42 (CTBP1-DT, CTBP1 Divergent Transcript) is an RNA Gene, and is affiliated with the non-coding RNA class.Specifications
C4orf42 | |
Monoclonal | |
Unconjugated | |
CTBP1-DT | |
CTBP1-DT | |
Human recombit protein fragment corresponding to amino acids 1-186 of human C4orf42 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI2C6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
C4orf42, CTBP1-AS1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
92070 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.