Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C4orf45 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570992
Description
C4orf45 Polyclonal Antibody for Western Blot
C4orf45 is a protein coding gene.
Specifications
C4orf45 | |
Polyclonal | |
Unconjugated | |
C4orf45 | |
C4orf45; chromosome 4 open reading frame 45; uncharacterized protein C4orf45 | |
Rabbit | |
Affinity chromatography | |
RUO | |
152940 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96LM5 | |
C4orf45 | |
synthetic peptide directed towards the C-terminal region of human C4orf45. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction