Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C4orf46 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561486
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561486 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561486 Supplier Invitrogen™ Supplier No. PA561486
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QVEELAFKCT ENARFLKTWR DLLKEGYDSL KPDD Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

This gene encodes a small, conserved protein of unknown function that is expressed in a variety of tissues. There are pseudogenes for this gene on chromosomes 6, 8, 16, and X. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Feb 2013].
TRUSTED_SUSTAINABILITY

Specifications

Antigen C4orf46
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C4orf46
Gene Accession No. Q504U0
Gene Alias 4930579G24Rik; C4orf46; chromosome 4 open reading frame 46; RCDG1; Renal cancer differentiation gene 1 protein; RIKEN cDNA 4930579G24 gene; uncharacterized protein C4orf46 homolog
Gene Symbols C4orf46
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C4orf46. Recombinant protein control fragment (Product #RP-100888).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201725
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.