Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C4orf46 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983546
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3546 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3546 Supplier Bioss Supplier No. BS15194R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a small, conserved protein of unknown function that is expressed in a variety of tissues. There are pseudogenes for this gene on chromosomes 6, 8, 16, and X. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Feb 2013].

Specifications

Antigen C4orf46
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene C4orf46
Gene Accession No. Q504U0
Gene Alias 4930579G24Rik; C4orf46; chromosome 4 open reading frame 46; RCDG1; Renal cancer differentiation gene 1 protein; RIKEN cDNA 4930579G24 gene; Uncharacterized protein C4orf46 homolog
Gene Symbols C4orf46
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C4orf46.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201725
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.