Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C4orf54 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5145200
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145200 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5145200 Supplier Invitrogen™ Supplier No. PA5145200
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FNRNEWKRKS DPLPMMMDSH VLSLIASEER EGVVVADGDH DKLSKRLGEV EERGTGNKAG VVLRGAPIER LQRRNSNPS Antigen sequence identity: Mouse 81%; Rat 29%.

C4orf54 is a protein that is encoded by the C4orf54 gene in humans. It is a poorly characterized protein with unknown function, but it has been suggested to play a role in the regulation of cell growth and differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C4orf54
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C4orf54
Gene Accession No. D6RIA3
Gene Alias Familial Obliterative Portal Venopathy; FOPV; LOC285556
Gene Symbols C4orf54
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range 1270-1348. Recombinant protein control fragment (Product #RP-110112).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285556
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.