Learn More
C5 Goat anti-Human, FITC, Polyclonal, Nordic

Goat Polyclonal Antibody
Supplier: Vector Laboratories GAHU/C5/FITC

Description
Dilutions may be prepared by adding phosphate buffered saline (PBS, pH 7.2). Repeated thawing and freezing should be avoided. If a slight precipitation occurs upon storage, this should be removed by centrifugation. It will not affect the performance of the antiserum. Diluted antiserum should be stored at 4 C, not refrozen, and preferably used the same day. In immunoelectrophoresis use 2 ul serum, plasma or equivalent against 120 ul antiserum. In double radial immunodiffusion (Ouchterlony) use a rosette arrangement with 10 ul antiserum in a 3 mm diameter centre well and 2 ul serum samples (neat and serially diluted) in 2 mm diameter peripheral wells. In single radial immunodiffusion and electroimmunodiffusion use 0.5 to 1.0 percent antiserum in the agar gel.
C5 Polyclonal antibody specifically detects C5 in Human samples. It is validated for Dot Blot, ELISA, Immunoblot, Immunocytochemistry, Immunohistochemistry (Frozen)Specifications
C5 | |
Polyclonal | |
FITC | |
C5 | |
anaphylatoxin; anaphylatoxin C5; anaphylatoxin C5a analog; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 4; C5; C5a; C5a anaphylatoxin; C5b; C5D; Complement C5; Complement C5 alpha chain; Complement C5 alpha' chain; Complement C5 beta chain; complement component 5; CPAMD4; ECLZB; Hc; He; Hemolytic complement; prepro-C5; RGD1561905 | |
Human C5 | |
1 mL | |
Primary | |
Human | |
Antibody | |
IgG |
Dot Blot, ELISA, Immunoblot, Immunocytochemistry, Immunohistochemistry (Frozen) | |
5-10 mg/mL | |
PBS with no preservative; pH | |
P01031 | |
Goat | |
Protein G | |
RUO | |
727 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.