Learn More
Sigma Aldrich C5 Lenalidomide-Dipiperazine-Nh2 Hydrochloride
Catalog No. 91989650MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 mg
Catalog No. Quantity
91-989-650MG 50 mg
1 options
Catalog No. 91-989-650MG Supplier Sigma Aldrich Supplier No. 91989650MG
Only null left
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Protein degrader building block Thalidomide-Photoswitch3-NH2 hydrochloride enables the synthesis of PHOtochemically TArgeting Chimeras (PHOTACs), or photoswitchable proteolysis-targeting chimeras (PROTACs) that can be reversibly activated with different wavelengths of light. Developed in the Trauner and Pagano labs, PHOTACs are inactive in the dark but are activated to the cis isomer via irradiation at 390 nm and reversibly deactivated at wavelengths above 450 nm. As described in Reynders et al, this conjugate was used to prepare PHOTAC-I-10 and PHOTAC-II-6 and contains a Cereblon (CRBN)-recruiting ligand, an azobenzene photoswitchable crosslinker, and a pendant amine for reactivity with an acid on the target warhead. Light-mediated control of the resulting PHOTAC affords advanced temporal and spatial control of targeted protein degradation. Suggested wavelengths for photoswitching:
  • Switch to cis isomer: 390 nm (380-400 nm)
  • Switch to trans isomer (thermally more stable isomer): >450 nm. Low-intensity light needed for photoactivation is not cytotoxic. Browse our full offering of degrader building blocks that streamlines the synthesis of degrader libraries.
Learn more: Technology Spotlight: Degrader Building Blocks for Targeted Protein DegradationPortal: Building PROTAC Degraders for Targeted Protein DegradationPHOTACs enable optical control of protein degradationPhotoPROTACs: A Novel Biotechnology for Cancer TreatmentPROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license
Specifications
CAS 9375-5-4-0
Quantity 50 mg
Synonym 2-(2-((6-Aminohexyl)oxy)ethoxy)-N-(2-(2,6-dioxopiperidin-3-yl)-1-oxoisoindolin-5-yl)acetamide hydrochloride; C5 Lenalidomide conjugate; Crosslinker-E3 Ligase ligand conjugate; Protein degrader building block for PROTAC(R) research; Template for synthesis of targeted protein degrader