Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C5 Mouse anti-Human, Clone: 2G8C9, Proteintech
SDP

Catalog No. 501737052 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7052 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7052 Supplier Proteintech Group Inc Supplier No. 666341IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C5 is part of the membrane attack complex (MAC) of the complement system. C5 is split into C5a og C5b by the C5 convertases from the classical and the alternative complement pathway and the C5b fragment initiates the assembly of the terminal MAC (C5b-C9). The molecular mass of C5 is 190 kDa.

Specifications

Antigen C5
Applications Western Blot
Classification Monoclonal
Clone 2G8C9
Concentration 0.93 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene C5
Gene Accession No. P01031
Gene Alias C5, Complement C5, complement component 5, CPAMD4
Gene Symbols C5
Host Species Mouse
Immunogen C5 Fusion Protein Ag17882
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 727
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.