Learn More
Invitrogen™ C5AR1 Monoclonal Antibody (20/70), PE
Rat Monoclonal Antibody
Supplier: Invitrogen™ MA181884
Description
Clone 20/70 has been reported to block the binding of the C5a to murine CD88. Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4°C in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Rat anti Mouse CD88 antibody, clone 20/70 recognizes murine CD88, also known as C5a anaphylatoxin chemotactic receptor 1 (C5aR1), a member of the G-protein coupled receptor 1 family.
CD88, also known as C5aR1, is a C5a Anaphylatoxin Receptor that mediates the effects of one of the most potent chemotactic agents known. C5AR1 has been implicated in asthma, and its expression has been reported in myeloid blood cells, brain, liver, lung, spleen, heart, kidney, and intestinal tract. CD88 is the receptor for the chemotactic and inflammatory peptide anaphylatoxin C5a. Functionally, CD88 stimulates chemotaxis, granule enzyme release and superoxide anion production. CD88 is also a component of the classical pathway of the complement cascade; it may be involved with tissue injury associated with ischemia or sepsis and it may be a component of the membrane attack complex. Diseases associated with CD88 include Mannose-Binding Lectin Deficiency and Systemic Lupus Erythematosus.
Specifications
C5AR1 | |
Monoclonal | |
Conc. Not Determined | |
PBS with 1% BSA, 5% sucrose and 0.09% sodium azide | |
P30993 | |
C5AR1 | |
RBL-2H3 transfected cells expressing murine C5aR. | |
100 Tests | |
Primary | |
Mouse | |
Antibody | |
IgG2b |
Flow Cytometry | |
20/70 | |
PE | |
C5AR1 | |
anaphylatoxin C5a receptor; C5A; C5a anaphylatoxin chemotactic receptor; C5a anaphylatoxin chemotactic receptor 1; C5a anaphylatoxin receptor; C5a ligand; C5a Receptor; C5AR; C5a-R; C5AR1; C5r1; Cd88; CD88 antigen; complement C5a receptor; complement C5a receptor 1; complement component 5 receptor 1; complement component 5, receptor 1; complement component 5a receptor 1; D7Msu1 | |
Rat | |
Protein G | |
RUO | |
12273 | |
4°C, store in dark, DO NOT FREEZE! | |
Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.