Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C5AR1 Recombinant Mouse Monoclonal Antibody (32-G1)
GREENER_CHOICE

Catalog No. PIMA548289
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA548289 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA548289 Supplier Invitrogen™ Supplier No. MA548289
Add to Cart
Add to Cart

Mouse Recombinant Monoclonal Antibody

Specificity: The 32-G1 antibody recognizes C5aR which has its serine 334 phosphorylated. The mAb 32-G1 reacted specifically with a C5aR peptide that contained phosphoserine 334 and also reacted with C5aRs that were isolated from C5a-stimulated, transfected RBL-C5aR cells. It did not recognize a receptor mutant (C5aR-S--) with alanine replacement of all six C-terminal serine phosphorylation sites. Antibody 32-G1 specifically reacts with C5aR phosphoserine 334, independent of the phosphorylation state of the neighboring serine residues. Studies with Bt2cAMP-treated U937 cells show that 32-G1 specifically reacts with ligand-activated C5aR at endogenous receptor expression levels but only weakly recognizes C5aR isolated from unstimulated cells.

CD88, also known as C5aR1, is a C5a Anaphylatoxin Receptor that mediates the effects of one of the most potent chemotactic agents known. C5AR1 has been implicated in asthma, and its expression has been reported in myeloid blood cells, brain, liver, lung, spleen, heart, kidney, and intestinal tract. CD88 is the receptor for the chemotactic and inflammatory peptide anaphylatoxin C5a. Functionally, CD88 stimulates chemotaxis, granule enzyme release and superoxide anion production. CD88 is also a component of the classical pathway of the complement cascade; it may be involved with tissue injury associated with ischemia or sepsis and it may be a component of the membrane attack complex. Diseases associated with CD88 include Mannose-Binding Lectin Deficiency and Systemic Lupus Erythematosus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C5AR1
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 32-G1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene C5AR1
Gene Accession No. P21730
Gene Alias anaphylatoxin C5a receptor; C5A; C5a anaphylatoxin chemotactic receptor; C5a anaphylatoxin chemotactic receptor 1; C5a anaphylatoxin receptor; C5a ligand; C5a Receptor; C5AR; C5a-R; C5AR1; C5r1; Cd88; CD88 antigen; complement C5a receptor; complement C5a receptor 1; complement component 5 receptor 1; complement component 5, receptor 1; complement component 5a receptor 1; D7Msu1
Gene Symbols C5AR1
Host Species Mouse
Immunogen This antibody was raised by immunizing mice a synthetic C5aR C-terminal phosphopeptide that contained phosphoserine instead of serine at position 334 and also at the neighboring phosphorylation sites Ser-332 and Ser-338.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 728
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.