Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C5b-9 Antibody (aE11) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 4 publications
Supplier: Novus Biologicals NBP1051200.025MG
Description
C5b-9 Monoclonal specifically detects C5b-9 in Human, Porcine, Bacteria, Equine, Baboon samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Western Blot (Negative).Specifications
C5b-9 | |
Monoclonal | |
1.0 mg/mL | |
ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Western Blot (Negative) | |
MAC, Membrane attack complex, sC5-b9, sC5b-9, TCC, Terminal complement complex | |
Purified C5b-9 | |
0.025 mg | |
Primary | |
Equine, Baboon, Pig, Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), ELISA, Immunofluorescence, Immunocytochemistry | |
aE11 | |
Unconjugated | |
10mM Phosphate (pH 7.4) and 0.5M NaCl with 0.05% Sodium Azide | |
Mouse | |
Protein A or G purified | |
RUO | |
Binds both membrane-bound MAC and fluid phase SC5b-9 complexes. | |
Store at 4C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction