Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C5b-9 Antibody (aE11), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP105120MFV610
Description
C5b-9 Monoclonal antibody specifically detects C5b-9 in Human, Porcine, Bacteria, Equine, Baboon samples. It is validated for Flow CytometrySpecifications
C5b-9 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Human, Pig, Bacteria, Equine, Baboon | |
Purified |
Flow Cytometry | |
aE11 | |
Flow Cytometry | |
MAC, Membrane attack complex, sC5-b9, sC5b-9, TCC, Terminal complement complex | |
Purified C5b-9 | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction