Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C5L2/GPR77 Antibody (468705), PE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB4729P
Description
C5L2/GPR77 Monoclonal antibody specifically detects C5L2/GPR77 in Mouse samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOFSpecifications
C5L2/GPR77 | |
Monoclonal | |
PE | |
PBS | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
27202 | |
Store at 4°C in the dark. | |
IgG2b |
Flow Cytometry (Intracellular Staining), CyTOF | |
468705 | |
Intracellular Staining by Flow Cytometry, CyTOF-ready | |
C5a anaphylatoxin chemotactic receptor C5L2, C5L2G-protein coupled receptor 77, G protein-coupled receptor 77, G protein-coupled receptor C5L2, GPF77 | |
Chinese hamster ovary cell line transfected with mouse C5L2, Met1-Val344, Accession # Q8BW93 | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction