Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C5L2/GPR77 Antibody (650918) [Allophycocyanin], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB10254A
Description
C5L2/GPR77 Monoclonal antibody specifically detects C5L2/GPR77 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
C5L2/GPR77 | |
Monoclonal | |
APC | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
27202 | |
Store at 4°C in the dark. | |
IgG2b |
Flow Cytometry, CyTOF | |
650918 | |
Flow Cytometry, CyTOF-ready | |
C5a anaphylatoxin chemotactic receptor C5L2, C5L2G-protein coupled receptor 77, G protein-coupled receptor 77, G protein-coupled receptor C5L2, GPF77 | |
NS0 mouse myeloma cell line transfected with human C5L2/GPR77, Met1-Val337, Accession # Q9P296 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction