Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C5orf24 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297988MFV610
Description
C5orf24 Polyclonal antibody specifically detects C5orf24 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
C5orf24 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
chromosome 5 open reading frame 24, FLJ37562, hypothetical protein LOC134553 | |
Produced in rabbits immunized with E. coli-derived Human C5orf24 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
134553 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction