Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C5orf24 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen C5orf24
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP152861
SDP
View Documents
Novus Biologicals
NBP152861
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

C5orf24 Polyclonal specifically detects C5orf24 in Human samples. It is validated for Western Blot.
Specifications

Specifications

C5orf24
Polyclonal
Rabbit
Q7Z6I8
134553
Synthetic peptides corresponding to C5ORF24 The peptide sequence was selected from the middle region of C5orf24. Peptide sequence SIEIKDELKKKKNLNRSGKRGRPSGTTKSAGYRTSTGRPLGTTKAAGFKT.
Primary
Western Blot
Unconjugated
RUO
chromosome 5 open reading frame 24, FLJ37562, hypothetical protein LOC134553
C5ORF24
IgG
20 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.