Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C5orf26 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5104050
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104050 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104050 Supplier Thermo Scientific Supplier No. PA5104050

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TIGA1.

C5orf26 (EPB41L4A Antisense RNA 1) is an RNA Gene, and is affiliated with the non-coding RNA class. Diseases associated with EPB41L4A-AS1 include Adamantinoma Of Long Bones.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C5orf26
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene EPB41L4A-AS1
Gene Accession No. 0
Gene Alias C5orf26; EPB41L4A antisense RNA 1; EPB41L4A-AS1; NCRNA00219; TIGA1
Gene Symbols EPB41L4A-AS1
Host Species Rabbit
Immunogen A synthesized peptide derived from human TIGA1, corresponding to a region within the internal amino acids.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114915
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.