Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C5orf28 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566297
Description
Immunogen sequence: VADRLLQFSTI QQNDWLRALS DNA Highest antigen sequence identity to the following orthologs - mouse 75%, rat 79%.
TMEM267 is a protein coding gene.
Specifications
C5orf28 | |
Polyclonal | |
Unconjugated | |
TMEM267 | |
C5orf28; TMEM267; Transmembrane protein 267; transmembrane protein C5orf28 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64417 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q0VDI3 | |
TMEM267 | |
Recombinant Human C5orf28. Recombinant protein control fragment (Product #RP-106978). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction