Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C5orf33 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558358
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558358 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558358 Supplier Invitrogen™ Supplier No. PA558358
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RELVEKVTNE YNESLLYSPE EPKILFSIRE PIANRVFSSS RQRCFSSKVC VRSRCWDACM VVDGGTSFEF NDGAIASMMI NKEDELRTVL LE Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 98%.

Mitochondrial NAD(+) kinase that phosphorylates NAD(+) to yield NADP(+). Can use both ATP or inorganic polyphosphate as the phosphoryl donor. Also has weak NADH kinase activity in vitro; however NADH kinase activity is much weaker than the NAD(+) kinase activity and may not be relevant in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C5orf33
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NADK2
Gene Accession No. Q4G0N4
Gene Alias C5orf33; DECRD; Mitochondrial NAD kinase; MNADK; NAD kinase 2, mitochondrial; NAD kinase domain containing 1; NAD kinase domain-containing protein 1, mitochondrial; NADK2; Nadkd1; RGD1306809
Gene Symbols NADK2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C5orf33. Recombinant protein control fragment (Product #RP-104107).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 133686
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.