Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C5orf33 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558359
Description
Immunogen sequence: RALNIERAHD ERSEASGPQL LPVRALNEVF IGESLSSRAS YYEISVDDGP WEKQKSSGLN LCTGTGSKAW SFNINRVATQ AVEDVLNIAK RQGNLSL Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 94%.
Mitochondrial NAD(+) kinase that phosphorylates NAD(+) to yield NADP(+). Can use both ATP or inorganic polyphosphate as the phosphoryl donor. Also has weak NADH kinase activity in vitro; however NADH kinase activity is much weaker than the NAD(+) kinase activity and may not be relevant in vivo.
Specifications
C5orf33 | |
Polyclonal | |
Unconjugated | |
NADK2 | |
C5orf33; DECRD; Mitochondrial NAD kinase; MNADK; NAD kinase 2, mitochondrial; NAD kinase domain containing 1; NAD kinase domain-containing protein 1, mitochondrial; NADK2; Nadkd1; RGD1306809 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
133686, 365699, 68646 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q1HCL7, Q4G0N4, Q8C5H8 | |
NADK2 | |
Recombinant protein corresponding to Human C5orf33. Recombinant protein control fragment (Product #RP-96901). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction