Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C5orf33 Polyclonal Antibody
C5orf33 Polyclonal Antibody
Supplier: Thermo Scientific PA598469
Description
C5orf33 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Specifications
C5orf33 | |
Polyclonal | |
Unconjugated | |
NADK2 | |
C5orf33; DECRD; Mitochondrial NAD kinase; MNADK; NAD kinase 2, mitochondrial; NAD kinase domain containing 1; NAD kinase domain-containing protein 1, mitochondrial; NADK2; Nadkd1; RGD1306809 | |
Rabbit | |
Protein G | |
RUO | |
133686, 365699, 68646 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
3.955 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q1HCL7, Q4G0N4, Q8C5H8 | |
NADK2 | |
Recombinant Human NAD kinase 2, mitochondrial protein (16-156AA). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction