Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C5orf34 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560990
Description
Immunogen sequence: LSLSHNYRIC CWKMVPGIND SNILPLVLKE SLIPSVGRFL AYSDDKVHAI FLDGITLTLN WNFSSPIEKR QVNQGLNLGW CKLTFPDGQE QLIQIEHPEP Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 76%.

Specifications
C5orf34 | |
Polyclonal | |
Unconjugated | |
C5orf34 | |
C5orf34; chromosome 5 open reading frame 34; uncharacterized protein C5orf34 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
375444 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96MH7 | |
C5orf34 | |
Recombinant protein corresponding to Human C5orf34. Recombinant protein control fragment (Product #RP-100232). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction