Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C5orf35 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113187
Description
C5orf35 Polyclonal Antibody for Western Blot, ELISA
SETD9 gene ontology annotations related to this gene include regulation of signal transduction by p53 class mediator.
Specifications
| C5orf35 | |
| Polyclonal | |
| Unconjugated | |
| SETD9 | |
| C5orf35; SET domain containing 9; SET domain-containing protein 9; SETD9 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 133383 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 3 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8NE22 | |
| SETD9 | |
| Recombinant Human SET domain-containing protein 9 protein (1-299AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction