Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C5ORF4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571426
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571426 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571426 Supplier Invitrogen™ Supplier No. PA571426
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C5ORF4 Polyclonal Antibody for Western Blot

FAXDC2 gene ontology annotations related to this gene include sterol biosynthetic process.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C5ORF4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FAXDC2
Gene Accession No. Q96IV6
Gene Alias C4H5orf4; C5orf4; fatty acid hydroxylase domain containing 2; fatty acid hydroxylase domain-containing protein 2; Faxdc2; uncharacterized protein C5orf4 homolog
Gene Symbols FAXDC2
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human C5ORF4.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10826
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.