Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C5ORF4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989100
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9100 Supplier Bioss Supplier No. BS9482R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAXDC2 gene ontology annotations related to this gene include sterol biosynthetic process.

Specifications

Antigen C5ORF4
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene FAXDC2
Gene Accession No. Q96IV6
Gene Alias C4H5orf4; C5orf4; chromosome 5 open reading frame 4-like; fatty acid hydroxylase domain containing 2; Fatty acid hydroxylase domain-containing protein 2; FAXDC2; LOC100347389
Gene Symbols FAXDC2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C5ORF4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10826
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.