Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C5orf4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP1602520 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP1602520 20 μL
NBP160125 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP1602520 Supplier Novus Biologicals Supplier No. NBP16012520UL

Rabbit Polyclonal Antibody

C5orf4 Polyclonal specifically detects C5orf4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen C5orf4
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS & 2% Sucrose. with No Preservative
Gene Alias chromosome 5 open reading frame 4, FLJ13758, hypothetical protein LOC10826
Gene Symbols C5ORF4
Host Species Rabbit
Immunogen Synthetic peptides corresponding to C5ORF4 The peptide sequence was selected from the N terminal of C5ORF4. Peptide sequence MKGEAGHMLHNEKSKQEGHIWGSMRRTAFILGSGLLSFVAFWNSVTWHLQ.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 10826
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.