Learn More
C5orf44 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | C5orf44 |
---|---|
Concentration | 0.37 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-2893
![]() ![]() |
Proteintech Group Inc
204441AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The MW of this protein is 47 kDa, and this antibody specifically recognizes the 47 kDa protein.
The function of C5orf44 has not been widely studied, and is yet to be fully elucidated.Specifications
C5orf44 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
A5PLN9, Q3TIR1, Q5M887 | |
294709, 66975, 80006 | |
C5orf44 Fusion Protein Ag14267 | |
Primary | |
-20°C | |
TRAPPC13 |
0.37 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
C5orf44, UPF0533 protein C5orf44 | |
Trappc13 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.