Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C5orf45 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113022
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113022 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113022 Supplier Invitrogen™ Supplier No. PA5113022
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C5orf45 Polyclonal Antibody for Western Blot, ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen C5orf45
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene MRNIP
Gene Accession No. Q6NTE8, Q9D1F5
Gene Alias 2900016G09Rik; 3010026O09Rik; AI851514; AW475975; C5orf45; chromosome 5 open reading frame 45; MRN complex-interacting protein; MRN-interacting protein; MRNIP; RGD1564036; RIKEN cDNA 3010026O09 gene; similar to RIKEN cDNA 3010026O09; UPF0544 protein C5orf45; UPF0544 protein C5orf45 homolog
Gene Symbols MRNIP
Host Species Rabbit
Immunogen Recombinant Human MRN complex-interacting protein (1-200AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51149, 68067
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.