Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C5orf49 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298031AF488
Description
C5orf49 Polyclonal specifically detects C5orf49 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
C5orf49 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
C5orf49 chromosome 5 open reading frame 49 | |
Produced in rabbits immunized with E. coli-derived Human C5orf49 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
134121 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction