Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C5orf54 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532073
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532073 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532073 Supplier Invitrogen™ Supplier No. PA532073
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: H1299, MCF-7. Predicted reactivity: Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

ZBED8 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C5orf54
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene ZBED8
Gene Accession No. Q8IZ13
Gene Alias Buster3; C5orf54; Protein ZBED8; transposon-derived Buster3 transposase-like protein; ZBED8; Zinc finger BED domain-containing protein 8; zinc finger BED-type containing 8; zinc finger, BED-type containing 8
Gene Symbols ZBED8
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human C5orf54. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63920
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.