Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C5orf54 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983561
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3561 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3561 Supplier Bioss Supplier No. BS15210R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZBED8 is a protein coding gene.

Specifications

Antigen C5orf54
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ZBED8
Gene Accession No. Q8IZ13
Gene Alias Buster3; C5orf54; Protein ZBED8; Transposon-derived Buster3 transposase-like protein; ZBED8; Zinc finger BED domain-containing protein 8; zinc finger BED-type containing 8; zinc finger, BED-type containing 8
Gene Symbols ZBED8
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C5orf54.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63920
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.