Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C5orf55 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569853
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569853 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569853 Supplier Invitrogen™ Supplier No. PA569853
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Human: 100%.

EXOC3-AS1 (EXOC3 Antisense RNA 1) is an RNA Gene, and is affiliated with the lncRNA class. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen C5orf55
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene EXOC3-AS1
Gene Accession No. Q8N2X6
Gene Alias C5orf55; EXOC3 antisense RNA 1; EXOC3-AS1
Gene Symbols EXOC3-AS1
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human C5orf55.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116349
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.