Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C5orf56 Monoclonal Antibody (OTI4B11), TrueMAB™, OriGene
SDP

Catalog No. 501684091
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-4091 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-4091 Supplier OriGene Supplier No. TA811508
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C5orf56 (Chromosome 5 Open Reading Frame 56) is a Protein Coding gene.

Specifications

Antigen C5orf56
Applications Western Blot
Classification Monoclonal
Clone OTI4B11
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene C5orf56
Gene Accession No. Q8N8D9
Gene Alias chromosome 5 open reading frame 56
Gene Symbols C5orf56
Host Species Mouse
Immunogen Full length human recombit protein of human C5orf56 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 441108
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.