Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C5orf60 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306034B
Description
C5orf60 Polyclonal specifically detects C5orf60 in Human samples. It is validated for Western Blot.Specifications
C5orf60 | |
Polyclonal | |
Western Blot | |
Chromosome 5 Open Reading Frame 60, Putative Uncharacterized Protein C5orf60, Putative Uncharacterized Protein FLJ35723 | |
Produced in rabbits immunized with E. coli-derived Human C5orf60 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Biotin | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
285679 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction