Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C5orf64 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544396
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544396 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544396 Supplier Invitrogen™ Supplier No. PA544396
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MLAPLFLCCL RNLFRKLISF QPPQLGRTNM HYSKLPRTAI ETEFKQNVGP Sequence homology: Human: 100%.

The specific function of the protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C5orf64
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene C5orf64
Gene Accession No. Q2M2E5
Gene Alias C5orf64; chromosome 5 open reading frame 64; uncharacterized protein C5orf64
Gene Symbols C5orf64
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human FLJ37543 (aa 1-50).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285668
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.