Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117369
Description
Centrifuge briefly prior to opening.
C6 is a component of complement cascade. It is part of the membrane attack complex which can insert into the cell membrane and cause cell to lyse. People with C6 deficiency are prone to bacterial infection. Two transcript variants encoding the same protein have been found for this gene.
Specifications
C6 | |
Polyclonal | |
Unconjugated | |
C6 | |
AW111623; C6; complement component 6; complement component C6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12274, 729 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
2.66 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P13671 | |
C6 | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human C6. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction