Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C6orf1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546354
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546354 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546354 Supplier Invitrogen™ Supplier No. PA546354
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SCSLTWETPR WYMAGRVATS TSGCHCWMSR RDLTPLPHPS EPGVLDCLGP Sequence homology: Human: 100%; Mouse: 85%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C6orf1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SMIM29
Gene Accession No. Q86T20
Gene Alias C6orf1; chromosome 6 open reading frame 1; LBH; Protein LBH; Small integral membrane protein 29; SMIM29; uncharacterized protein C6orf1
Gene Symbols SMIM29
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human C6orf1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221491
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.