Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C6orf1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558046
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558046 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558046 Supplier Invitrogen™ Supplier No. PA558046
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LRNCMRLSRS CSLTWETPRW YMAGRVATST SGCHCWMSRR DLTPLPHPSE PGVLDCLGPC HLLPLLSP Highest antigen sequence identity to the following orthologs: Mouse - 26%, Rat - 34%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C6orf1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SMIM29
Gene Accession No. Q86T20
Gene Alias C6orf1; chromosome 6 open reading frame 1; LBH; Protein LBH; Small integral membrane protein 29; SMIM29; uncharacterized protein C6orf1
Gene Symbols SMIM29
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C6orf1. Recombinant protein control fragment (Product #RP-95027).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221491
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.