Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C6orf1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121469
This item is not returnable.
View return policy
Description
Antibody tested in Indirect ELISA.
The function of this protein remains unknown.
Specifications
C6orf1 | |
Polyclonal | |
Unconjugated | |
SMIM29 | |
C6orf1; chromosome 6 open reading frame 1; LBH; Protein LBH; Small integral membrane protein 29; SMIM29; uncharacterized protein C6orf1 | |
Rabbit | |
Protein G | |
RUO | |
221491 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry | |
4.02 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q86T20 | |
SMIM29 | |
Recombinant Human Uncharacterized protein SMIM29 protein (42-102aa). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction