Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C6orf106 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983564
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3564 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3564 Supplier Bioss Supplier No. BS15213R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Potential guanine nucleotide exchange factor (GEF). GEF proteins activate some small GTPases by exchanging bound GDP for free GTP.

Specifications

Antigen C6orf106
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene C6orf106
Gene Accession No. Q9H6K1
Gene Alias AU020189; C030010A15; C6orf106; C80239; chromosome 6 open reading frame 106; D17Wsu92e; dJ391O22.4; DNA segment, Chr 17, Wayne State University 92, expressed; FP852; ILRUN; Inflammation and lipid regulator with UBA-like and NBR1-like domains protein; Protein ILRUN; uncharacterized protein C6orf106; uncharacterized protein C6orf106 homolog
Gene Symbols C6orf106
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C6orf106.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64771
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.