Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C6orf115 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556401
Description
Immunogen sequence: NVDHEVNLLV EEIHRLGSKN ADGKLSVKFG VLFRDDKCAN LFEALVGTLK AAKRRKIVTY PGELLLQGVH DDVDIILLQD Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 94%.
Specifications
| C6orf115 | |
| Polyclonal | |
| Unconjugated | |
| ABRACL | |
| 1700028I04Rik; 3110003A17Rik; ABRA C-terminal like; ABRA C-terminal-like protein; Abracl; C6orf115; Costars; Costars family protein ABRACL; HSPC280; PRO2013 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 58527, 685045, 73112 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q4KML4, Q9P1F3 | |
| ABRACL | |
| Recombinant protein corresponding to Human C6orf115. Recombinant protein control fragment (Product #RP-94894). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction