Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C6orf120 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546380
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546380 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546380 Supplier Invitrogen™ Supplier No. PA546380
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: KVYYDGTVEQ HPFGEAAYPA DGADAGQKHA GAPEDASQEE ESVLWTILIS Sequence homology: Cow: 92%; Human: 100%; Rat: 86%.

C6orf120 may be involved in induction of apoptosis in CD4(+) T- cells, but not CD8(+) T-cells or hepatocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C6orf120
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene C6orf120
Gene Accession No. Q7Z4R8
Gene Alias bA160E12.4; C6orf120; chromosome 6 open reading frame 120; UPF0669 protein C6orf120
Gene Symbols C6orf120
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human C6orf120 (aa 132-181).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387263
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.