Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C6orf129 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560953
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560953 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560953 Supplier Invitrogen™ Supplier No. PA560953
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KKRENLGVAL EIDGLEEKLS QCRRDLEAVN SRLHSRELS Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 85%.

CCDC167 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C6orf129
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCDC167
Gene Accession No. Q9P0B6
Gene Alias 1110021J02Rik; C23H6orf129; C6orf129; CCDC167; coiled-coil domain containing 167; coiled-coil domain-containing protein 167; HSPC265; transmembrane and coiled-coil domain-containing protein C6orf129; transmembrane and coiled-coil domain-containing protein C6orf129 homolog
Gene Symbols CCDC167
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C6orf129. Recombinant protein control fragment (Product #RP-100013).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 154467
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.