Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C6orf136 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306307PE
Description
C6orf136 Polyclonal antibody specifically detects C6orf136 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
C6orf136 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
chromosome 6 open reading frame 136, Em:AB023049.8, hypothetical protein LOC221545, MGC15854 | |
Produced in rabbits immunized with E. coli-derived Human C6orf136 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
221545 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction