Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C6orf136 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS15219R

Description
C6orf136 is a protein coding gene.Specifications
C6orf136 | |
Polyclonal | |
Unconjugated | |
C6orf136 | |
C6orf136; chromosome 6 open reading frame 136; Uncharacterized protein C6orf136 | |
Rabbit | |
Protein A | |
RUO | |
221545 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q5SQH8 | |
C6orf136 | |
KLH conjugated synthetic peptide derived from human C6orf136. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction