Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C6orf145 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983573
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3573 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3573 Supplier Bioss Supplier No. BS15222R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PXDC1 gene ontology annotations related to this gene include phosphatidylinositol binding.

Specifications

Antigen C6orf145
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PXDC1
Gene Accession No. Q5TGL8
Gene Alias C6orf145; PX domain containing 1; PX domain-containing protein 1; PX domain-containing protein C6orf145; PXDC1
Gene Symbols PXDC1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C6orf145.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221749
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.