Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C6orf150 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501092719 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-109-2719 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-109-2719 Supplier Proteintech Group Inc Supplier No. 264161AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The exact function of C6orf150 remains unknown.

Specifications

Antigen C6orf150
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene CGAS
Gene Accession No. Q8N884
Gene Alias C6orf150, cGAS, Cyclic GMP-AMP synthase, h-cGAS, MB21D1
Gene Symbols CGAS
Host Species Rabbit
Immunogen C6orf150 Fusion Protein Ag23455
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115004
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.